missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-USP13, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 500-550 of human USP13 was used as the immunogen for the antibody.
Antigen USP13
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP224575SS Novus Biologicals Canada
NBP224575SS
0.025mg Each for $109.00
NBP224575 Novus Biologicals Canada
NBP224575
0.1mg Each for $257.41
Specifications Description & Specifications

Specifications

Rabbit
USP13
Affinity Purified
RUO
This antibody is reactive to Chimpanzee, Cow, Dog, Horse, Human, Mouse, Rhesus Monkey.
8975
0.5mg/mL
A portion of amino acids 500-550 of human USP13 was used as the immunogen for the antibody.
Polyclonal
Primary
Immunohistochemistry (Paraffin),Western Blot
Q92995
100μg antibody in 200μL PBS containing 0.05% BSA