Please login to your online account to display your discounted pricing

anti-USP13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224575SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224575SS

  • / Each
 

Description & Specifications

Specifications

Antigen USP13
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Equine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Gene Accession No. Q92995
Host Species Rabbit
Immunogen A portion of amino acids 500-550 of human USP13 was used as the immunogen for the antibody.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 8975
Test Specificity This antibody is reactive to Chimpanzee, Cow, Dog, Horse, Human, Mouse, Rhesus Monkey.
Target Species Bovine
Target Species Canine
Target Species Equine
Target Species Human
Target Species Murine
Target Species Primate