Please login to your online account to display your discounted pricing

anti-ULK2, Polyclonal, Novus Biologicals™

$160.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 693 and 968 of ULK2 (Uniprot ID No.Q8IYT8)
Antigen ULK2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP133136

 
novus biologicals canada
NBP133136
Unlabeled 0.1mg Each for $384.00

NBP133136SS

 
novus biologicals canada
NBP133136SS
Unlabeled 0.02mg Each for $160.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Recombinant fragment corresponding to a region within amino acids 693 and 968 of ULK2 (Uniprot ID No.Q8IYT8)
Antigen ULK2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity Not yet tested in any other species.
Gene ID (Entrez) 9706
Formulation 0.1M Tris, 0.1M Glycine, 10% Glycerol (pH 7.0).
Concentration 1.0mg/mL