Please login to your online account to display your discounted pricing

anti-ULK2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP133136SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP133136SS

  • / Each
 

Description & Specifications

Specifications

Immunogen Recombinant fragment corresponding to a region within amino acids 693 and 968 of ULK2 (Uniprot ID No.Q8IYT8)
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 0.02mg
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene ID (Entrez) 9706
Concentration 1.0mg/mL
Conjugate Unlabeled
Primary or Secondary Primary
Formulation 0.1M Tris, 0.1M Glycine, 10% Glycerol (pH 7.0).
Monoclonal or Polyclonal Polyclonal
Test Specificity Not yet tested in any other species.
Regulatory Status RUO
Antigen ULK2
Host Species Rabbit
Format Affinity Purified