Please login to your online account to display your discounted pricing

anti-Uev1a/UBE2V1, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 31-49 of human UBE2V1a. Genbank accession no. NP_954595.1.
Antigen Uev1a/UBE2V1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056397

 
novus biologicals canada
NB10056397
Unlabeled 0.1mg Each for $255.80

NB10056397T

 
novus biologicals canada
NB10056397SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 31-49 of human UBE2V1a. Genbank accession no. NP_954595.1.
Antigen Uev1a/UBE2V1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity A protein band of approximate mloecular weght of 20 kD can be seen in Jurkat, RAW and HeLa cell lysates. This may also recognize isoform b of UBE2V1. Cross reacts with Human. Not yet tested in other species.
Gene ID (Entrez) 7335
Formulation PBS containing 0.2% gelatin
Concentration 0.5mg/mL