Please login to your online account to display your discounted pricing

anti-Uev1a/UBE2V1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056397SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056397T

  • / Each
 

Description & Specifications

Specifications

Antigen Uev1a/UBE2V1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.2% gelatin
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 31-49 of human UBE2V1a. Genbank accession no. NP_954595.1.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7335
Test Specificity A protein band of approximate mloecular weght of 20 kD can be seen in Jurkat, RAW and HeLa cell lysates. This may also recognize isoform b of UBE2V1. Cross reacts with Human. Not yet tested in other species.
Target Species Human