Please login to your online account to display your discounted pricing

anti-UCP4, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human UCP4 was used as the immunogen for the antibody.
Antigen UCP4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224562SS

 
novus biologicals canada
NBP224562SS
Unlabeled 0.025mg Each for $139.00

NBP224562

 
novus biologicals canada
NBP224562
Unlabeled 0.1mg Each for $259.02
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human UCP4 was used as the immunogen for the antibody.
Antigen UCP4
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat and mouse, and 91% homologous in pig and bovine.
Gene ID (Entrez) 9481
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL