Please login to your online account to display your discounted pricing

anti-UCP4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224562SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224562SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9481
Antigen UCP4
Format Affinity Purified
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen A portion of amino acids 250-300 of human UCP4 was used as the immunogen for the antibody.
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat and mouse, and 91% homologous in pig and bovine.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Quantity 0.025mg