Please login to your online account to display your discounted pricing

anti-UCP4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224562

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224562

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 9481
Primary or Secondary Primary
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, rat and mouse, and 91% homologous in pig and bovine.
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Host Species Rabbit
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Porcine
Cross Reactivity Primate
Cross Reactivity Rat
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Conjugate Unlabeled
Format Affinity Purified
Immunogen A portion of amino acids 250-300 of human UCP4 was used as the immunogen for the antibody.
Antigen UCP4
Quantity 0.1mg