Please login to your online account to display your discounted pricing

anti-UCP3, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 125-175 of human UCP3 was used as the immunogen.
Antigen UCP3
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224608

 
novus biologicals canada
NBP224608
Unlabeled 0.1mg Each for $307.00

NBP224608SS

 
novus biologicals canada
NBP224608SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 125-175 of human UCP3 was used as the immunogen.
Antigen UCP3
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human and chimpanzee, 94% homologous in horse, wild boar, macaque, rhesus monkey, cow and dog, 88% homologous in mouse and rat, and 83% homologous in opossum.
Gene Accession No. P55916
Gene ID (Entrez) 7352
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL