Please login to your online account to display your discounted pricing

anti-Ubiquitin, Polyclonal, Novus Biologicals™

$139.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Rabbits were immunized with purified ubiquitin conjugated to keyhole limpet hemocyanin.
Antigen Ubiquitin
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP230132

 
novus biologicals canada
NBP230132
Unlabeled 0.1mL Each for $384.00

NBP230132SS

 
novus biologicals canada
NBP230132SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Rabbits were immunized with purified ubiquitin conjugated to keyhole limpet hemocyanin.
Antigen Ubiquitin
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Test Specificity Cat, Human, New World monkey, Most Mammals, Mouse, Rat.
Gene Accession No. NP_001129064
Gene ID (Entrez) 6233
Formulation 50% glycerol, 1.0mg/mL BSA.