Please login to your online account to display your discounted pricing

anti-Ubiquitin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP230132SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP230132SS

  • / Each
 

Description & Specifications

Specifications

Formulation 50% glycerol, 1.0mg/mL BSA.
Gene Accession No. NP_001129064
Test Specificity Cat, Human, New World monkey, Most Mammals, Mouse, Rat.
Gene ID (Entrez) 6233
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Quantity 0.025mL
Antigen Ubiquitin
Primary or Secondary Primary
Format Antiserum
Immunogen Rabbits were immunized with purified ubiquitin conjugated to keyhole limpet hemocyanin.
Host Species Rabbit
Regulatory Status RUO