Please login to your online account to display your discounted pricing

anti-Ubiquitin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP230132

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP230132

  • / Each
 

Description & Specifications

Specifications

Antigen Ubiquitin
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Feline
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Antisera
Formulation 50% glycerol, 1.0mg/mL BSA.
Gene Accession No. NP_001129064
Host Species Rabbit
Immunogen Rabbits were immunized with purified ubiquitin conjugated to keyhole limpet hemocyanin.
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 6233
Test Specificity Cat, Human, New World monkey, Most Mammals, Mouse, Rat.
Target Species Feline
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat