Please login to your online account to display your discounted pricing

anti-UBE2N/Ubc13, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides containing amino acids 2-19 and 131-148 of human UBE2N.
Antigen UBE2N/Ubc13
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056357T

 
novus biologicals canada
NB10056357SS
Unlabeled 0.025mg Each for $139.00

NB10056357

 
novus biologicals canada
NB10056357
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides containing amino acids 2-19 and 131-148 of human UBE2N.
Antigen UBE2N/Ubc13
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Western Blotting
Test Specificity UBE2N .
Gene Accession No. P61088
Gene ID (Entrez) 7334
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL