Please login to your online account to display your discounted pricing

anti-UBE2N/Ubc13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056357SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056357T

  • / Each
 

Description & Specifications

Specifications

Antigen UBE2N/Ubc13
Applications Immunoprecipitation
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. P61088
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides containing amino acids 2-19 and 131-148 of human UBE2N.
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7334
Test Specificity UBE2N .
Target Species Human