Please login to your online account to display your discounted pricing

anti-UBE2N/Ubc13, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056357

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056357

  • / Each
 

Description & Specifications

Specifications

Formulation PBS containing 0.05% BSA
Gene Accession No. P61088
Host Species Rabbit
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7334
Test Specificity UBE2N .
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen UBE2N/Ubc13
Format Purified
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides containing amino acids 2-19 and 131-148 of human UBE2N.