Please login to your online account to display your discounted pricing

anti-UbcH6/UBE2E1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP178983SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP178983SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Gene Accession No. P51965
Gene ID (Entrez) 7324
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Concentration 1.02mg/mL
Quantity 0.025mL
Formulation PBS, 30% glycerol
Host Species Rabbit
Regulatory Status RUO
Antigen UbcH6/UBE2E1
Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Immunogen A synthetic peptide made to an N-terminal portion of the human UBCH6 protein (between residues 1-50) [UniProt P51965]
Format Affinity Purified