Please login to your online account to display your discounted pricing

anti-Tyrosine Kinase Receptor B Polyclonal, Thermo Scientific™

Tyrosine Kinase Receptor B Polyclonal Ab PA1-18403 from Thermo Scientific™ detects Tyrosine Kinase Receptor B in Ms and Rt samples, and is validated for ELISA and IHC

$509.25 - $522.90

Specifications

Antigen Tyrosine Kinase Receptor B
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-652

 
pierce biotechnology inc
PA118402
Unlabeled 100μL Each for $509.25

01-670-653

 
pierce biotechnology inc
PA118403
Unlabeled 100μL Each for $522.90
Description & Specifications

Specifications

Antigen Tyrosine Kinase Receptor B
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Extracellular domain of glycosylated mouse TrkB protein produced in CHO cells.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Tyrosine Kinase Receptor B Polyclonal Ab PA1-18403 from Thermo Scientific™ detects Tyrosine Kinase Receptor B in Ms and Rt samples, and is validated for ELISA and IHC

This gene encodes a member of the neurotrophic tyrosine receptor kinase (NTRK) family. This kinase is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signalling through this kinase leads to cell differentiation. Mutations in this gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for this gene.
Host Species: Rabbit
Cross Reactivity: Mouse, Rat
Isotype: IgG
Immunogen: Extracellular domain of glycosylated mouse TrkB protein produced in CHO cells.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: TrkB
Uniprot Accession: P15209, Q63604
Gene Symbol: Ntrk2
Entrez GeneID: 18212, 25054
PA1-18403 detects Tyrosine Kinase Receptor B in Mouse and Rat samples. PA1-18403 has been successfully used in ELISA and Immunohistochemistry procedures. PA1-18403 The immunogen is Extracellular domain of glycosylated mouse TrkB protein produced in CHO cells..