Please login to your online account to display your discounted pricing

anti-Tyrosine Hydroxylase Polyclonal, Thermo Scientific™

Tyrosine Hydroxylase Polyclonal Ab PA1-18315 from Thermo Scientific™ specifically detects Tyrosine Hydroxylase in Gp, Ms and Rt samples, and is validated for IHC (P,F)

$506.05 - $506.05

Specifications

Antigen Tyrosine Hydroxylase
Cross Reactivity Guinea Pig
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-568

 
pierce biotechnology inc
PA118315
Unlabeled 100μL Each for $506.05
Description & Specifications

Specifications

Antigen Tyrosine Hydroxylase
Cross Reactivity Guinea Pig
Cross Reactivity Mouse
Cross Reactivity Rat
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues P (63) R F I G R R Q S L I E D A R K(78) of human Tyrosine Hydroxylase conjugated to KLH.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Tyrosine Hydroxylase Polyclonal Ab PA1-18315 from Thermo Scientific™ specifically detects Tyrosine Hydroxylase in Gp, Ms and Rt samples, and is validated for IHC (P,F)

catalyzes the rate limiting step in catecholamine biosynthesis; may be involved in response to hypoxia.
Host Species: Rabbit
Cross Reactivity: Guinea Pig, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues P (63) R F I G R R Q S L I E D A R K(78) of human Tyrosine Hydroxylase conjugated to KLH.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: TH, Tyrosine 3-monooxygenase, L-tyrosine hydroxylase, Tyrosine 3-hydroxylase
Uniprot Accession: P04177, P24529
Gene Symbol: Th
Entrez GeneID: 100734150, 21823, 25085
PA1-18315 detects Tyrosine Hydroxylase in Guinea Pig, Mouse and Rat samples. PA1-18315 has been successfully used in Immunohistochemistry (Paraffin and Frozen) procedures. PA1-18315 The immunogen is Synthetic peptide corresponding to residues P (63) R F I G R R Q S L I E D A R K(78) of human Tyrosine Hydroxylase conjugated to KLH..