Please login to your online account to display your discounted pricing

anti-TWF2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 from human TWF2 was used as the immunogen for this antibody.
Antigen TWF2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227171

 
novus biologicals canada
NBP227171
Unlabeled 0.1mg Each for $255.80

NBP227171SS

 
novus biologicals canada
NBP227171SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 1-50 from human TWF2 was used as the immunogen for this antibody.
Antigen TWF2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The TWF2 amino acid sequence used as immunogen for this antibody is 100% homologous in human, new world monkey, rhesus monkey, orangutan, chimpanzee, 95% in cow, Chinese hamster, mouse, rat, and pig, 89% in panda, Carolina anole, dog, mole rat, elephant, opossum, platypus, and frog, 84% in zebrafish, horse, chicken, zebra finch, and 79% homologouus in cichlid fish.
Gene Accession No. Q6IBS0
Gene ID (Entrez) 11344
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Concentration 0.5mg/mL