Please login to your online account to display your discounted pricing

anti-TWF2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227171

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227171

  • / Each
 

Description & Specifications

Specifications

Antigen TWF2
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA.
Gene Accession No. Q6IBS0
Host Species Rabbit
Applications Western Blotting
Immunogen A portion of amino acids 1-50 from human TWF2 was used as the immunogen for this antibody.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 11344
Test Specificity The TWF2 amino acid sequence used as immunogen for this antibody is 100% homologous in human, new world monkey, rhesus monkey, orangutan, chimpanzee, 95% in cow, Chinese hamster, mouse, rat, and pig, 89% in panda, Carolina anole, dog, mole rat, elephant, opossum, platypus, and frog, 84% in zebrafish, horse, chicken, zebra finch, and 79% homologouus in cichlid fish.
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human