missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TWEAK R/TNFRSF12, Polyclonal, Novus Biologicals™

$139.00 - $255.80

Specifications

Host Species Rabbit
Immunogen This antibody was developed against a synthetic peptide containing amino acids 106-118 (CRREKFTTPIEETG) of human TweakR.
Antigen TWEAK R/TNFRSF12
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB10056636T Novus Biologicals Canada
NB10056636SS
0.025mg Each for $139.00
NB10056636 Novus Biologicals Canada
NB10056636
0.1mg Each for $255.80
Specifications Description & Specifications

Specifications

Rabbit
TWEAK R/TNFRSF12
Purified
RUO
51330
0.5mg/mL
This antibody was developed against a synthetic peptide containing amino acids 106-118 (CRREKFTTPIEETG) of human TweakR.
Polyclonal
Primary
Western Blot
PBS containing 0.05% BSA