missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
TUC-4 Protein Rabbit anti-Human, Mouse, Monkey, Rat, Feline, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB5454
Description
Anti-TUC-4 Protein Antibody detects level of TUC-4 Protein and has been published and validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.Specifications
TUC-4 Protein | |
Polyclonal | |
Rabbit Serum. Liquid containing 0.2% sodium azide. | |
Rabbit | |
Unpurified | |
Primary | |
Feline, Human, Monkey, Mouse, Rat | |
Serum |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
DPYSL3; CRMP-4; DRP3; CRMP4; ULIP; DRP-3 | |
Synthetic peptide corresponding to amino acids 499 to 511 of TUC-4a and amino acids 612-624 of TUC-4b (YDGPVFDLTTTPK) | |
100 μL | |
NM_001387.2 | |
−20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only