missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
tubulin, epsilon 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human TUBE1 protein.
Supplier: Abnova Corporation H00051175B01
Description
This gene encodes a member of the tubulin superfamily. This protein localizes to the centriolar sub-distal appendages that are associated with the older of the two centrioles after centrosome duplication. This protein plays a central role in organization of the microtubules during centriole duplication. A pseudogene of this gene is found on chromosome 5
Sequence: MTQSVVVQVGQCGNQIGCCFWDLALREHAAVNQKGIYDEAISSFFRNVDTRVVGDGGSISKGKICSLKARAVLIDMEEGVVNEILQGPLRDVFDTKQLITDISGSGNNWAVGHKVFGSLYQDQILEKFRKSAEHCDCLQCFFIIHSMGGGTGSGLGTFLLKVLEDEFPEVYRFVTSIYPSGEDDVITSPYNSILAMKELNEHADCVLPIDNQSLFDIISKIDLMVNSGKLGTTVKPKSLVTSSSGALKKQHKKPFDAMNNIVANLLLNLTSSARFEGSLNMDLNEISMNLVPFPQLHYLVSSLTPLYTLTDVNIPPRRLDQMFSDAFSKDHQLLRADPKHSLYLACALMVRGNVQISDLRRNIERLKPSLQFVSWNQEGWKTSLCSVPPVGHSHSLLALANNTCVKPTFMELKERFMRLYKKKAHLHHYLQVEGMEESCFTEAVSSLSALIQEYDQLDATKNMPVQDLPRLSIAMSpecifications
tubulin, epsilon 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human TUBE1 protein. | |
TUBE1 | |
FLJ22589/TUBE/dJ142L7.2 | |
Mouse | |
50 μL | |
Cell Cycle | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
NM_016262 | |
TUBE1 | |
TUBE1 (NP_057346, 1 a.a. ∼ 475 a.a) full-length human protein. | |
RUO | |
Yes | |
51175 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |