Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP227288

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP227288

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Conjugate Unlabeled
Formulation 100μL; TSLP receptor antibody sera with 50% glycerol.
Gene ID (Entrez) 64109
Immunogen Amino acids 145-178 of human TSLP receptor protein were used as the immunogen for the antibody.
Quantity 0.1mL
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Antigen TSLP R/CRLF2
Regulatory Status RUO
Format Antiserum
Applications Immunohistochemistry (Paraffin)
Host Species Rabbit