Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Murine
Isotype κ
Immunogen Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody.
Antigen TSLP R/CRLF2
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP222384

 
novus biologicals canada
NBP222384
Unlabeled 0.1mg Each for $307.00

NBP222384SS

 
novus biologicals canada
NBP222384SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Murine
Isotype κ
Immunogen Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody.
Antigen TSLP R/CRLF2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. AF338733_1
Gene ID (Entrez) 64109
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA