Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222384SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222384SS

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. AF338733_1
Isotype κ
Quantity 0.025mg
Gene ID (Entrez) 64109
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Conjugate Unlabeled
Format Purified
Immunogen Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody.
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Regulatory Status RUO
Cross Reactivity Human
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen TSLP R/CRLF2
Host Species Mouse