Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP222384

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP222384

  • / Each
 

Description & Specifications

Specifications

Antigen TSLP R/CRLF2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation 0.1mg antibody in 0.2mL PBS containing 0.05% BSA
Gene Accession No. AF338733_1
Host Species Murine
Immunogen Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody.
Isotype κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 64109
Target Species Human