Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen Partial recombinant human CRLF2 protein (between amino acids 100-300) [UniProt Q9HC73]
Antigen TSLP R/CRLF2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP233302SS

 
novus biologicals canada
NBP233302SS
Unlabeled 0.025mg Each for $109.00

NBP233302

 
novus biologicals canada
NBP233302
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Partial recombinant human CRLF2 protein (between amino acids 100-300) [UniProt Q9HC73]
Antigen TSLP R/CRLF2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Gene Accession No. Q9HC73
Gene ID (Entrez) 64109
Formulation PBS
Concentration 1.0mg/mL