Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 145-178 of human TSLP receptor protein were used as the immunogen for the antibody.
Antigen TSLP R/CRLF2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227288SS

 
novus biologicals canada
NBP227288SS
Unlabeled 0.025mL Each for $109.00

NBP227288

 
novus biologicals canada
NBP227288
Unlabeled 0.1mL Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 145-178 of human TSLP receptor protein were used as the immunogen for the antibody.
Antigen TSLP R/CRLF2
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Gene ID (Entrez) 64109
Formulation 100μL; TSLP receptor antibody sera with 50% glycerol.