Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233302SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233302SS

  • / Each
 

Description & Specifications

Specifications

Antigen TSLP R/CRLF2
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS
Gene Accession No. Q9HC73
Host Species Rabbit
Immunogen Partial recombinant human CRLF2 protein (between amino acids 100-300) [UniProt Q9HC73]
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 64109
Target Species Human