Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP233302

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP233302

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Conjugate Unlabeled
Format Purified
Formulation PBS
Gene Accession No. Q9HC73
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Immunogen Partial recombinant human CRLF2 protein (between amino acids 100-300) [UniProt Q9HC73]
Primary or Secondary Primary
Gene ID (Entrez) 64109
Concentration 1.0mg/mL
Cross Reactivity Human
Quantity 0.1mg
Antigen TSLP R/CRLF2
Regulatory Status RUO