Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229613SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229613SS

  • / Each
 

Description & Specifications

Specifications

Antigen TSLP R/CRLF2
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 1.0mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS
Gene Accession No. Q9HC73
Host Species Rabbit
Immunogen Partial recombinant protein made to an internal portion of human CRLF2 protein (between residues 100-300) [NCBI AF338733_1]
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 64109
Target Species Human