Please login to your online account to display your discounted pricing

anti-TSLP R/CRLF2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP229613

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP229613

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 64109
Concentration 1.0mg/mL
Gene Accession No. Q9HC73
Cross Reactivity Human
Format Purified
Conjugate Unlabeled
Formulation PBS
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Host Species Rabbit
Immunogen Partial recombinant protein made to an internal portion of human CRLF2 protein (between residues 100-300) [NCBI AF338733_1]
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Antigen TSLP R/CRLF2
Regulatory Status RUO
Quantity 0.1mg