Please login to your online account to display your discounted pricing

anti-TSLP, Polyclonal, Novus Biologicals™

$106.00 - $106.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human TSLP protein was used as the immunogen.
Antigen TSLP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP223664SS

 
novus biologicals canada
NBP223664SS
Unlabeled 0.025mg Each for $106.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 100-150 of human TSLP protein was used as the immunogen.
Antigen TSLP
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee, monkey and cow, 93% homologous in mouse and dog, 86% homologous in opossum, and 73% homologous in rat.
Gene ID (Entrez) 85480
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL