Please login to your online account to display your discounted pricing

anti-TSLP, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP223664SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP223664SS

  • / Each
 

Description & Specifications

Specifications

Antigen TSLP
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 85480
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chimpanzee, monkey and cow, 93% homologous in mouse and dog, 86% homologous in opossum, and 73% homologous in rat.
Concentration 0.5mg/mL
Conjugate Unlabeled
Immunogen A portion of amino acids 100-150 of human TSLP protein was used as the immunogen.
Quantity 0.025mg
Regulatory Status RUO
Cross Reactivity Human
Cross Reactivity Primate