Please login to your online account to display your discounted pricing

TSKS Polyclonal Antibody, Invitrogen™


TSKS Antibody, PA1-16660, from Invitrogen™. Species Reactivity: Human; Applications: ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

Manufacturer: pierce biotechnology inc  PA116660

Catalog No. APPA6660

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Formulation 100μL unpurified antibody in whole serum and 0.02% sodium azide
Gene ID (Entrez) 60385
Storage Requirements avoiding freeze/thaw cycles.
Gene Symbols TSKS
Species Reactivity Human
Gene Accession No. Q9UJT2
Format Serum
Gene Alias STK22S1, TSKS1, TSSKS
Quantity 100μL
Cross Reactivity Human
Isotype IgG
Primary or Secondary Primary
Conjugate Unlabeled
Monoclonal or Polyclonal Polyclonal
Antigen TSKS
Host Species Rabbit
Immunogen Residues 577-592 of the human TSKS protein

TSKS Antibody, PA1-16660, from Invitrogen. Species Reactivity: Human; Applications: ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot.

A novel putative serine/threonine kinase substrate gene (TSKS, for testis-specific kinase substrate) is likely the human homolog of the mouse testis-specific serine kinase substrate gene. TSKS exhibits high levels of expression exclusively in human testicular tissue. The expression of TSKS is downregulated in cancerous testicular tissue, in comparison to adjacent normal tissue. These observations suggest a role of TSKS in testicular physiology, most probably in the process of spermatogenesis or spermiogenesis.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Residues 577-592 of the human TSKS protein
Formulation: 100μL unpurified antibody in whole serum and 0.02% sodium azide
Synonyms: STK22S1, TSKS1, TSSKS
Uniprot Accession: Q9UJT2
Entrez GeneID: 60385
Gene Symbol: TSKS
This product detects TSKS in Human samples. This product has been successfully used in ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) and Western Blot procedures. The immunogen is Residues 577-592 of the human TSKS protein.