Please login to your online account to display your discounted pricing

anti-TSKS Polyclonal, Thermo Scientific™

TSKS Polyclonal Ab PA1-16660 from Thermo Scientific™ specifically detects TSKS in Hu samples, and it is validated for ELISA, IHC (Frozen), IHC(P) and WB

$448.05 - $448.05

Specifications

Antigen TSKS
Cross Reactivity Human
Format Serum
Host Species Rabbit
Immunogen Residues 577-592 of the human TSKS protein
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

APPA6660

 
pierce biotechnology inc
PA116660
Unlabeled 100μL Each for $448.05
Description & Specifications

Specifications

Antigen TSKS
Cross Reactivity Human
Format Serum
Host Species Rabbit
Immunogen Residues 577-592 of the human TSKS protein
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements avoiding freeze/thaw cycles.

TSKS Polyclonal Ab PA1-16660 from Thermo Scientific™ specifically detects TSKS in Hu samples, and it is validated for ELISA, IHC (Frozen), IHC(P) and WB

A novel putative serine/threonine kinase substrate gene (TSKS, for testis-specific kinase substrate) is likely the human homolog of the mouse testis-specific serine kinase substrate gene. TSKS exhibits high levels of expression exclusively in human testicular tissue. The expression of TSKS is downregulated in cancerous testicular tissue, in comparison to adjacent normal tissue. These observations suggest a role of TSKS in testicular physiology, most probably in the process of spermatogenesis or spermiogenesis.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Residues 577-592 of the human TSKS protein
Formulation: 100μL unpurified Antibody in whole serum, with 0.02% sodium azide;
Synonyms: STK22S1, TSKS1, TSSKS
Uniprot Accession: Q9UJT2
Gene Symbol: TSKS
Entrez GeneID: 60385
PA1-16660 detects TSKS in Human samples. PA1-16660 has been successfully used in ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) and Western Blot procedures. PA1-16660 The immunogen is Residues 577-592 of the human TSKS protein.

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly