Please login to your online account to display your discounted pricing

anti-Tryptophan hydroxylase 2, Polyclonal, Novus Biologicals™

$109.00 - $259.02

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 1-100) [UniProt Q8IWU9]
Antigen Tryptophan hydroxylase 2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP177556

 
novus biologicals canada
NBP177556
Unlabeled 0.1mL Each for $259.02

NBP177556SS

 
novus biologicals canada
NBP177556SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 1-100) [UniProt Q8IWU9]
Antigen Tryptophan hydroxylase 2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene Accession No. Q8IWU9
Gene ID (Entrez) 121278
Formulation PBS, 30% glycerol
Concentration 1.19mg/mL