Please login to your online account to display your discounted pricing

anti-Tryptophan hydroxylase 2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177556SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177556SS

  • / Each
 

Description & Specifications

Specifications

Antigen Tryptophan hydroxylase 2
Applications Western Blot
Concentration 1.19mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q8IWU9
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of the human Tryptophan hydroxylase 2 protein (between residues 1-100) [UniProt Q8IWU9]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 121278
Target Species Human