Please login to your online account to display your discounted pricing

anti-TRPM7, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB500243

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB500243

  • / Each
 

Description & Specifications

Specifications

Antigen TRPM7
Format Antiserum
Host Species Rabbit
Applications Immunoprecipitation
Applications Western Blotting
Immunogen A synthetic peptide made to the C-terminal region of rat TRPM7. [UniProt No. Q925B3]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 54822
Test Specificity NB 500-243 recognizes overexpressed human TRPM7. It does not appear to recognize overexpressed mouse TRPM7. It does not work on endogenous samples tested for western analysis (human, rat and mouse cell lines. Other species have not been tested.
Conjugate Unlabeled
Cross Reactivity Human