Please login to your online account to display your discounted pricing

anti-TRPM2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081601

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081601

  • / Each
 

Description & Specifications

Specifications

Monoclonal or Polyclonal Polyclonal
Concentration 0.90mg/mL
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Conjugate Unlabeled
Formulation PBS, 30% glycerol
Regulatory Status RUO
Cross Reactivity Mouse
Cross Reactivity Rat
Quantity 0.1mL
Primary or Secondary Primary
Gene ID (Entrez) 7226
Test Specificity This is specific for isoform 1, or the long isoform (TRPM2-L), of TRPM2.
Antigen TRPM2
Immunogen Synthetic peptide made to a C-terminal portion of the rat TRPM2 protein (within residues 1430-1508). [Swiss-Prot No. Q5G856]
Host Species Rabbit
Format Affinity Purified