missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TRPM2, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the rat TRPM2 protein (within residues 1430-1508). [Swiss-Prot No. Q5G856]
Antigen TRPM2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NB11081601 Novus Biologicals Canada
NB11081601
0.1mL Each for $329.00
NB11081601T Novus Biologicals Canada
NB11081601SS
0.025mL Each for $139.00
Specifications Description & Specifications

Specifications

Rabbit
TRPM2
Affinity Purified
RUO
This is specific for isoform 1, or the long isoform (TRPM2-L), of TRPM2.
PBS, 30% glycerol
Synthetic peptide made to a C-terminal portion of the rat TRPM2 protein (within residues 1430-1508). [Swiss-Prot No. Q5G856]
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Frozen),Western Blot
7226
0.90mg/mL