Please login to your online account to display your discounted pricing

anti-TRPM2, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081601SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081601T

  • / Each
 

Description & Specifications

Specifications

Antigen TRPM2
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blot
Concentration 0.90mg/mL
Conjugate Unlabeled
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation PBS, 30% glycerol
Host Species Rabbit
Immunogen Synthetic peptide made to a C-terminal portion of the rat TRPM2 protein (within residues 1430-1508). [Swiss-Prot No. Q5G856]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 7226
Test Specificity This is specific for isoform 1, or the long isoform (TRPM2-L), of TRPM2.
Target Species Murine
Target Species Rat