Please login to your online account to display your discounted pricing

anti-TROY/TNFRSF19, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB12012126SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB12012126T

  • / Each
 

Description & Specifications

Specifications

Antigen TROY/TNFRSF19
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. Q9NS68
Host Species Rabbit
Immunogen 204-219
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 55504
Test Specificity Cross-reacts with Human and Mouse. Not yet tested in other species.
Target Species Human
Target Species Murine