missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

anti-TrkC, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the extracellular domain of the human TrkC protein (within residues 300-400). [UniProt Q16288].
Antigen TrkC
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP215185 Novus Biologicals Canada
NBP215185
0.1mL Each for $309.05
NBP215185SS Novus Biologicals Canada
NBP215185SS
0.025mL Each for $109.00
Specifications Description & Specifications

Specifications

Rabbit
TrkC
Affinity Purified
RUO
Human and mouse. Immunogen has 100% homology to pig and monkey.
4916
1.16mg/mL
A synthetic peptide made to the extracellular domain of the human TrkC protein (within residues 300-400). [UniProt Q16288].
Polyclonal
Primary
Immunocytochemistry,Immunofluorescence,Immunohistochemistry,Immunohistochemistry (Paraffin)
Q16288
PBS, 30% glycerol