Trimethyl-Histone H4 (Lys20) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer: Emd Millipore Corporation 07463
Description
Anti-trimethyl-Histone H4 (Lys20) Antibody is a rabbit polyclonal antibody for detection of trimethyl-Histone H4 (Lys20) also known as H4K20me3, Histone H4 (tri methyl K20) and has been validated in immunoprecipitation, western blot, Chimmunoprecipitation, DB.Specifications
trimethyl Histone H4 (Lys20) | |
Unconjugated | |
Affinity Purified | |
Rabbit | |
Affinity purified | |
2-8°C for 12 months | |
Polyclonal | |
Human,Murine,Rat |
Chromatin Immunoprecipitation,Dot Blot,Immunoprecipitation,Western Blot | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HIST1H4A; H4/J; HIST1H4F; HIST1H4L; H4FN; H4FA; H4FH; HIST1H4H; H4F2; H4FM; H4/A; H4FK; H4FG; HIST2H4; H4FB; H4/K; HIST1H4I; H4/N; HIST1H4B; H4FE; H4/M; H4/a; HIST1H4E; HIST4H4; HIST2H4A; H4FC; H4FI; H4/H; HIST1H4C; H4/G; HIST1H4K; H4FJ; H4FD; H4/I; H4/B; H4/C; H4/D; HIST1H4J; HIST1H4D; H4/E | |
KLH-conjugated linear peptide corresponding to the N-terminus of human Histone H4 trimethylated at Lys20. | |
100μg | |
Primary | |
NP_001029249 |
For Research Use Only