Please login to your online account to display your discounted pricing

anti-trimethyl-Histone H3 (Lys9), Clone: 6F12-H4, EMD Millipore™


Manufacturer: emd millipore corporation  051242

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. 051242MI

  • / Each
 

Description & Specifications

Specifications

Storage Requirements 2-8°C for one year. Avoid freezing, which can damage IgG and affect product performance.
Isotype κ
Immunogen KLH-conjugated, branched synthetic peptide containing the sequence (QTARme3K- STGGKA)2-KC, in which me3K corresponds to trimethyl lysine 9 of human histone H3. Data suggests this antibody may require the presence of the glutamine residue in position 5 fo
Conjugate Unlabeled
Applications Chromatin Immunoprecipitation
Applications Dot Blot
Applications Immunofluorescence
Applications Peptide Inhibition Assay
Applications Western Blotting
Antigen trimethyl-Histone H3 (Lys9)
Formulation Purified antibody supplied at 1mg/mL in 0.1MTris-glycine, pH 7.4, 150mM NaCl, 0.05% NaN3.
Purification Method Purified
Host Species Mouse
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Quantity 200μL
Gene Symbols H3.4, H3/g, H3/t, H3FT, H3t, MGC126886, OTTHUMP00000037945
Clone 6F12-H4
Gene Accession No. NM_003493
Format Purified
Monoclonal or Polyclonal Monoclonal