missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethyl Histone H3 (Lys79) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$175.14 - $697.80
Specifications
Antigen | trimethyl Histone H3 (Lys79) |
---|---|
Applications | Multiplex, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
07-952-SMI
|
MilliporeSigma
07952S |
10 μg |
Each for $175.14
|
|
|||||
07-952-MI
|
MilliporeSigma
07952 |
100 μg |
Each for $697.80
|
|
|||||
Description
Anti-trimethyl Histone H3 (Lys79) Antibody is a rabbit polyclonal antibody for detection of trimethyl Histone H3 (Lys79) also known as H3K79me2, Histone H3 (di methyl K79) and has been validated in western blot, Mplex.Specifications
trimethyl Histone H3 (Lys79) | |
Polyclonal | |
Rabbit | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
HIST3H3; H3FT; H3t; H3T; H3/g; H3.4; H3/t | |
Primary | |
2-8°C for 12 months |
Multiplex, Western Blot | |
Unconjugated | |
Human | |
NP_003484.1 | |
Linear Peptide corresponding to human Histone H3 methylated at Lys79. | |
Affinity Purified |
For Research Use Only