missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Trimethyl Histone H3 (Lys79) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer: Emd Millipore Corporation 07952S
Description
Anti-trimethyl Histone H3 (Lys79) Antibody is a rabbit polyclonal antibody for detection of trimethyl Histone H3 (Lys79) also known as H3K79me2, Histone H3 (di methyl K79) and has been validated in western blot, Mplex.Specifications
trimethyl Histone H3 (Lys79) | |
Unconjugated | |
Affinity Purified | |
Rabbit | |
Affinity Purfied | |
2-8°C for 12 months | |
Polyclonal | |
Human |
Immunoassay,Multiplexing,Western Blot | |
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide. | |
HIST3H3; H3FT; H3t; H3T; H3/g; H3.4; H3/t | |
Linear Peptide corresponding to human Histone H3 methylated at Lys79. | |
10μg | |
Primary | |
NP_003484.1 |
For Research Use Only